CH6371380787 Chem Impex International, Inc.

Boc-O-benzyl-L-b-homohydroxyproline dicyclohexylammonium salt

  • MDL number MFCD03788173
  • Cas Number : 336182-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-beta-HomoHyp(Bzl)-OH DCHA
Purity 98
Storage temp. Store at 0-8
Physical Form White to off- white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-l-beta-homohydroxyproline-dicyclohexylammonium-salt/5909?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-l-beta-homohydroxyproline-dicyclohexylammonium-salt/5909?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart