CH6371380797 Chem Impex International, Inc.

Z-L-methionine methyl ester

  • MDL number MFCD00145551
  • Cas Number : 56762-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-43
Synonym Z-L-Met-OMe
Purity 98
Storage temp. Store at 0-8
Physical Form Light yellowish oil
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-methionine-methyl-ester/5921?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-methionine-methyl-ester/5921?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart