CH6371380803 Chem Impex International, Inc.

Boc-5-isothiocyanatopentylamine

  • MDL number MFCD01862960
  • Cas Number : 347890-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Tert-Butyl N-(5-isothiocyanatopentyl)carbamate
Purity 0.99
Density 1.05
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-5-isothiocyanatopentylamine/5928?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-5-isothiocyanatopentylamine/5928?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart