CH6371380806 Chem Impex International, Inc.

Boc-2-isothiocyanatoethylamine

  • MDL number MFCD00674499
  • Cas Number : 137743-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-89 °C
Purity 95
Storage temp. Store at 0-8 °C
Physical Form Pale yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-2-isothiocyanatoethylamine/5933?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-2-isothiocyanatoethylamine/5933?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart