CH6371380812 Chem Impex International, Inc.

Fmoc-2-methyl-D-phenylalanine

  • MDL number MFCD00671404
  • Cas Number : 352351-63-4
  • Molecular Weight : 401.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110 - 117 °C
Purity 99.5
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-2-methyl-d-phenylalanine/5939?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-2-methyl-d-phenylalanine/5939?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart