CH6371380821 Chem Impex International, Inc.

Boc-2-cyano-L-phenylalanine

  • MDL number MFCD01860623
  • Cas Number : 216312-53-7
  • Molecular Weight : 290.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-97 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-2-cyano-l-phenylalanine/5949?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-2-cyano-l-phenylalanine/5949?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart