CH6371380879 Chem Impex International, Inc.

L-Alanine allyl ester hydrochloride

  • MDL number MFCD01074682
  • Cas Number : 203799-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Ala-OAll HCl
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-alanine-allyl-ester-hydrochloride/6039?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-alanine-allyl-ester-hydrochloride/6039?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart