CH6371380887 Chem Impex International, Inc.

N1-Boc-N4-Fmoc-piperazine-2-carboxylic acid

  • MDL number MFCD01076217
  • Cas Number : 218278-58-1
  • Molecular Weight : 452.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/n-sup-1-sup-boc-n-sup-4-sup-fmoc-piperazine-2-carboxylic-acid/6048?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-sup-1-sup-boc-n-sup-4-sup-fmoc-piperazine-2-carboxylic-acid/6048?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart