CH6371380895 Chem Impex International, Inc.

beta-Cyclopentyl-D-alanine

  • MDL number MFCD03788182
  • Cas Number : 99295-81-5
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/ß-cyclopentyl-d-alanine/6057?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ß-cyclopentyl-d-alanine/6057?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart