CH6371380898 Chem Impex International, Inc.

Boc-L-Phe(2-trifluoromethyl)-OH

  • MDL number MFCD01862946
  • Cas Number : 167993-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-phe-2-trifluoromethyl-oh/6060?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-phe-2-trifluoromethyl-oh/6060?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart