CH6371380905 Chem Impex International, Inc.

3-Amino-1-carboxymethyl-pyridin-2-one�

  • MDL number MFCD03788184
  • Cas Number : 1373519-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym ACPO-OH TFA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-amino-1-carboxymethyl-pyridin-2-one-font-size-2-face-symbol-middot-font-tfa/6068?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-amino-1-carboxymethyl-pyridin-2-one-font-size-2-face-symbol-middot-font-tfa/6068?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart