CH6371380909 Chem Impex International, Inc.

Na-Fmoc-D-2,3-diaminopropionic acid hydrochloride

  • MDL number MFCD03788624
  • Cas Number : 251317-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-187
Synonym Fmoc-D-Dap-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-d-2-3-diaminopropionic-acid-hydrochloride/6072?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-d-2-3-diaminopropionic-acid-hydrochloride/6072?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart