CH6371380912 Chem Impex International, Inc.

Nalpha-Boc-5-hydroxy-L-tryptophan

  • MDL number MFCD00270732
  • Cas Number : 119768-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-5-hydroxy-l-tryptophan/6075?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-5-hydroxy-l-tryptophan/6075?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart