CH6371380914 Chem Impex International, Inc.

Boc-4-nitro-L-phenylalanine dicyclohexylammonium salt

  • MDL number MFCD00069882
  • Cas Number : 102185-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-171
Synonym Boc-L-Phe(4-NO2)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-4-nitro-l-phenylalanine-dicyclohexylammonium-salt/6079?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-4-nitro-l-phenylalanine-dicyclohexylammonium-salt/6079?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart