CH6371380922 Chem Impex International, Inc.

Phg-Gly-OH

  • MDL number MFCD06796734
  • Cas Number : 134624-91-2
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238-242 °C
Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phg-gly-oh/6087?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/phg-gly-oh/6087?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart