CH6371380927 Chem Impex International, Inc.

Boc-D-phenylalanine methyl ester

  • MDL number MFCD00672509
  • Cas Number : 77119-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Phe-OMe
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-phenylalanine-methyl-ester/6096?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-phenylalanine-methyl-ester/6096?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart