CH6371380936 Chem Impex International, Inc.

(R)-1-Z-2-tert-butyl-3-methyl-4-imidazolidinone

  • MDL number MFCD00075099
  • Cas Number : 119906-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-85
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-1-z-2-tert-butyl-3-methyl-4-imidazolidinone/6106?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-1-z-2-tert-butyl-3-methyl-4-imidazolidinone/6106?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart