CH6371380937 Chem Impex International, Inc.

(R)-1-Boc-2-tert-butyl-3-methyl-4-imidazolidinone

  • MDL number MFCD00075100
  • Cas Number : 119838-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-70
Synonym (R)-(+)-1-(tert-Butoxycarbonyl)-2-tert-butyl-3-methyl-4-imidazolidinone
Purity 99
Storage temp. Store at 0-8
Physical Form White fine crystals with lumps
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-1-boc-2-i-tert-i-butyl-3-methyl-4-imidazolidinone/6107?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-1-boc-2-i-tert-i-butyl-3-methyl-4-imidazolidinone/6107?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart