CH6371380948 Chem Impex International, Inc.

L-2,4-Diaminobutyric acid hydrochloride

  • MDL number MFCD01632031
  • Cas Number : 1482-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217-226 �C
Synonym L-Dab-OH�
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-2-4-diaminobutyric-acid-hydrochloride/6123?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-2-4-diaminobutyric-acid-hydrochloride/6123?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart