CH6371380949 Chem Impex International, Inc.

L-3,4-Dichlorophenylalanine methyl ester hydrochloride

  • MDL number MFCD06796736
  • Cas Number : 173522-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167-169
Synonym L-Phe(3,4-DiCl)-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-3-4-dichlorophenylalanine-methyl-ester-hydrochloride/6124?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-3-4-dichlorophenylalanine-methyl-ester-hydrochloride/6124?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart