CH6371380954 Chem Impex International, Inc.

a-Methyl-L-phenylalanine methyl ester hydrochloride

  • MDL number MFCD06796741
  • Cas Number : 64619-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-118
Synonym alpha-Me-L-Phe-OMe HCl
Purity 97
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/alpha-methyl-l-phenylalanine-methyl-ester-hydrochloride/6131?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/alpha-methyl-l-phenylalanine-methyl-ester-hydrochloride/6131?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart