CH6371380967 Chem Impex International, Inc.

DL-Glutamic acid g-tert-butyl ester

  • MDL number MFCD08458515

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DL-Glu(OtBu)-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-glutamic-acid-i-tert-i-butyl-ester/6147?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-glutamic-acid-i-tert-i-butyl-ester/6147?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart