CH6371380969 Chem Impex International, Inc.

Na-Fmoc-Nim-benzyl-D-histidine

  • MDL number MFCD08458517
  • Cas Number : 2022956-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-His(Bzl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-im-sup-benzyl-d-histidine/6149?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-im-sup-benzyl-d-histidine/6149?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart