CH6371380971 Chem Impex International, Inc.

Pra-NH2�

  • MDL number MFCD09753266

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/pra-nh-sub-2-sub-font-size-2-face-symbol-middot-font-tfa/6151?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/pra-nh-sub-2-sub-font-size-2-face-symbol-middot-font-tfa/6151?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart