CH6371380972 Chem Impex International, Inc.

(S)-Acetamidomethyl-L- penicillamine hydrochloride

  • MDL number MFCD08458518
  • Cas Number : 1330286-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Pen(Acm)-OH HCl
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-acetamidomethyl-l-penicillamine-hydrochloride/6152?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-acetamidomethyl-l-penicillamine-hydrochloride/6152?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart