CH6371380976 Chem Impex International, Inc.

L-2-Naphthylalanine methyl ester hydrochloride

  • MDL number MFCD08458519
  • Cas Number : 63024-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-179
Synonym L-Ala(2-naphthyl)-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White to yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-2-naphthylalanine-methyl-ester-hydrochloride/6156?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-2-naphthylalanine-methyl-ester-hydrochloride/6156?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart