CH6371380981 Chem Impex International, Inc.

Spectinomycin sulfate

  • MDL number MFCD00270185
  • Cas Number : 23312-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/spectinomycin-sulfate/6162?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/spectinomycin-sulfate/6162?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart