CH6371380991 Chem Impex International, Inc.

3-Bromo-D-phenylalanine

  • MDL number MFCD06659100
  • Cas Number : 99295-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Phe(3-Br)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-bromo-d-phenylalanine/6173?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-bromo-d-phenylalanine/6173?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart