CH6371381000 Chem Impex International, Inc.

1,3,3-Trimethoxypropene

  • MDL number MFCD00041903
  • Cas Number : 17576-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.06
Storage temp. Store at RT
Physical Form Colorless liquid
Shipping Temperature RT
Country of Origin USA
Boiling point 70-76 °C @ 16mm
Product Link http://www.chemimpex.com/product/productinfo/1-3-3-trimethoxypropene/6184?cid=0

Safety Information

hcodes 3
MSDS http://www.chemimpex.com/product/productinfo/1-3-3-trimethoxypropene/6184?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart