CH6371381003 Chem Impex International, Inc.

3,5-Diiodo-D-tyrosine hydrochloride

  • MDL number MFCD08457603

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-192 °C (dec.)
Purity 99
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-5-diiodo-d-tyrosine-hydrochloride/6187?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-5-diiodo-d-tyrosine-hydrochloride/6187?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart