CH6371381010 Chem Impex International, Inc.

(S)-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD00037512
  • Cas Number : 22916-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195 - 199 °C
Purity 99
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-5-5-dimethyl-1-3-thiazolidine-4-carboxylic-acid/6196?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-5-5-dimethyl-1-3-thiazolidine-4-carboxylic-acid/6196?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart