CH6371381012 Chem Impex International, Inc.

Na-Boc-L-2,4-diaminobutyric acid tert-butyl ester oxalate

  • MDL number MFCD11113156

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-160
Synonym Boc-L-Dab-OtBu Oxalate
Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-l-2-4-diaminobutyric-acid-i-tert-i-butyl-ester-oxalate/6198?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-l-2-4-diaminobutyric-acid-i-tert-i-butyl-ester-oxalate/6198?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart