CH6371381019 Chem Impex International, Inc.

3-Chloro-D-tyrosine

  • MDL number MFCD09842592
  • Cas Number : 162599-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228-252 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-chloro-d-tyrosine/6206?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-chloro-d-tyrosine/6206?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart