CH6371381022 Chem Impex International, Inc.

D-Norleucine methyl ester hydrochloride

  • MDL number MFCD09263353
  • Cas Number : 60687-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Nle-OMe HCl
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-norleucine-methyl-ester-hydrochloride/6211?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-norleucine-methyl-ester-hydrochloride/6211?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart