CH6371381025 Chem Impex International, Inc.

2',3',5'-Triacetyl -azacytidine

  • MDL number MFCD11113159
  • Cas Number : 10302-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-3-5-triacetyl-azacytidine/6217?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-3-5-triacetyl-azacytidine/6217?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart