CH6371381027 Chem Impex International, Inc.

(+)-Biotin-sarcosine

  • MDL number MFCD11113162

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-184 �darkens @ 173
Synonym Biotin-Sarcosine
Purity 97
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/biotin-sarcosine/6221?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/biotin-sarcosine/6221?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart