CH6371381030 Chem Impex International, Inc.

Boc-O-benzyl-D-b-homoserine

  • MDL number MFCD08282798
  • Cas Number : 718608-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-beta-HomoSer(Bzl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-beta-homoserine/6226?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-beta-homoserine/6226?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart