CH6371381042 Chem Impex International, Inc.

Boc-S-allyloxy-amidomethyl-L-cysteine dicyclohexylamine salt

  • MDL number MFCD00273400

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Cys(Allocam)-OH DCHA
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-allyloxy-amidomethyl-l-cysteine-dicyclohexylamine-salt/6250?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-allyloxy-amidomethyl-l-cysteine-dicyclohexylamine-salt/6250?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart