CH6371381045 Chem Impex International, Inc.

(S)-Pyrrolidine-2-carboxylic acid 2-butoxyethyl ester trifluroacetate

  • MDL number MFCD00273390

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Pro-OBg TFA
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-pyrrolidine-2-carboxylic-acid-2-butoxyethyl-ester-trifluroacetate/6253?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-pyrrolidine-2-carboxylic-acid-2-butoxyethyl-ester-trifluroacetate/6253?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart