CH6371381046 Chem Impex International, Inc.

Boc-O-allyloxycarbonyl-L-serine dicyclohexylammonium salt

  • MDL number MFCD00273417

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-112
Synonym Boc-L-Ser(Aloc)-OH DCHA
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-allyloxycarbonyl-l-serine-dicyclohexylammonium-salt/6254?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-allyloxycarbonyl-l-serine-dicyclohexylammonium-salt/6254?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart