CH6371381047 Chem Impex International, Inc.

Boc-O-allyloxycarbonyl-L-threonine dicyclohexylammonium salt

  • MDL number MFCD00273419

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-133
Synonym Boc-L-Thr(Aloc)-OH DCHA
Purity 96
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-allyloxycarbonyl-l-threonine-dicyclohexylammonium-salt/6255?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-allyloxycarbonyl-l-threonine-dicyclohexylammonium-salt/6255?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart