CH6371381048 Chem Impex International, Inc.

L-Tryptophan allyl ester hydrochloride

  • MDL number MFCD01074685

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-215
Synonym L-Trp-OAll HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-tryptophan-allyl-ester-hydrochloride/6256?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-tryptophan-allyl-ester-hydrochloride/6256?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart