CH6371381050 Chem Impex International, Inc.

Fmoc-1-amino-cyclohexane acetic acid

  • MDL number MFCD01863220
  • Cas Number : 282524-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-130 °C
Purity 95
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-1-amino-cyclohexane-acetic-acid/6258?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-1-amino-cyclohexane-acetic-acid/6258?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart