CH6371381053 Chem Impex International, Inc.

Boc-cis-DL-4-phenylpiperidine-3-carboxylic acid

  • MDL number MFCD08457835

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-184
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-cis-dl-4-phenylpiperidine-3-carboxylic-acid/6261?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-cis-dl-4-phenylpiperidine-3-carboxylic-acid/6261?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart