CH6371381055 Chem Impex International, Inc.

(R)-Boc-4-amino-5-(3-indolyl)pentanoic acid

  • MDL number MFCD06410965

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-161
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-boc-4-amino-5-3-indolyl-pentanoic-acid/6263?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-boc-4-amino-5-3-indolyl-pentanoic-acid/6263?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart