CH6371381070 Chem Impex International, Inc.

(1R,2R)-Boc-aminocyclopentane carboxylic acid

  • MDL number MFCD04974223
  • Cas Number : 245115-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1R,2R)-Boc-Acpc
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-i-r-i-2-i-r-i-boc-aminocyclopentane-carboxylic-acid/6279?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-i-r-i-2-i-r-i-boc-aminocyclopentane-carboxylic-acid/6279?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart