CH6371381076 Chem Impex International, Inc.

Fmoc-3-aminophenylacetic acid

  • MDL number MFCD02682314
  • Cas Number : 186320-08-1
  • Molecular Weight : 373.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-198 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-aminophenylacetic-acid/6285?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-aminophenylacetic-acid/6285?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart