CH6371381077 Chem Impex International, Inc.

4-(Boc-4-aminophenyl)butanoic acid

  • MDL number MFCD06656431
  • Cas Number : 105300-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-109
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-boc-4-aminophenyl-butanoic-acid/6286?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-boc-4-aminophenyl-butanoic-acid/6286?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart