CH6371381082 Chem Impex International, Inc.

(R,S)-Boc-2-amino-tetradecanoic acid

  • MDL number MFCD07781256
  • Cas Number : 129850-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-63
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/r-s-boc-2-amino-tetradecanoic-acid/6291?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/r-s-boc-2-amino-tetradecanoic-acid/6291?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart