CH6371381090 Chem Impex International, Inc.

Boc-4-cyclohexyl-piperidine-4-carboxylic acid

  • MDL number MFCD08457834
  • Cas Number : 273378-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-4-cyclohexyl-piperidine-4-carboxylic-acid/6299?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-4-cyclohexyl-piperidine-4-carboxylic-acid/6299?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart